Learn More
Economy Food Calorimeter
Click to view available options
:
Economy Food Calorimeter
Description
Includes
- Chamber
- Flask
- Cork and pin
- Instructions
This calorimeter is designed specifically to determine the caloric content of various foodstuffs. A flask containing a known mass of water is suspended in the calorimeter chamber. A nut of known mass is placed beneath the flask and burned. Heat from the burned nut heats up the water in the flask and raises its temperature. Using the increase in water temperature, students calculate the caloric content of the food.
Specifications
Specifications
Product Type | Economy Food Calorimeter |
For Use With (Application) | Determine the heat energy stored in foods, students can calculate the caloric content of the food |
Includes | Chamber, flask, cork and pin, instructions |