Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECS Polyclonal Antibody, QED Bioscience™

Goat Polyclonal Antibody
$482.32 - $3247.60
Specifications
Antigen | ECS |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7833
![]() ![]() |
Leinco Technologies Inc
1883001100UG |
100 μg |
Each for $482.32
|
|
|||||
50-227-7834
![]() ![]() |
Leinco Technologies Inc
188301MG |
1 mg |
Each for $3,247.60
|
|
|||||
Description
ECS Polyclonal antibody specifically detects ECS in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
ECS | |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Goat | |
PBS with 0.09% sodium azide; pH 7.2 | |
DDDDK conjugated to KLH. | |
Protein A | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
DDDDK; ECS; ECS Cleavage Site | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title