Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECSIT Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB087207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087207 Supplier Novus Biologicals Supplier No. NBP300303AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECSIT Polyclonal specifically detects ECSIT in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ECSIT
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ECSIT (Q9BQ95-1; Glu246-Ser431).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51295
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.