Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECSIT Antibody, Novus Biologicals™
SDP

Catalog No. NBP176594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176594 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176594 Supplier Novus Biologicals Supplier No. NBP176594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECSIT Polyclonal specifically detects ECSIT in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ECSIT
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_057665
Gene Alias ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved
Gene Symbols ECSIT
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human ECSIT. The immunogen is located within amino acids 290 - 340 of ECSIT. Amino Acid Squence: PPEEREVEETPEEW
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 51295
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.