Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDA/Ectodysplasin Antibody (Renzo-2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019767 0.025 mg
NBP259772 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019767 Supplier Novus Biologicals Supplier No. NBP2597720.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.

Specifications

Antigen EDA/Ectodysplasin
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Renzo-2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:100
Formulation PBS with 0.02% Sodium Azide
Gene Alias ectodysplasin A, ED1
Gene Symbols EDA
Host Species Mouse
Immunogen Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391).
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1896
Test Specificity Recognizes human and mouse EDA-A1 and EDA-A2.
Target Species Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.