Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDA/Ectodysplasin Antibody (Renzo-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | EDA/Ectodysplasin |
---|---|
Clone | Renzo-2 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:100 |
Applications | ELISA, Flow Cytometry, Immunoprecipitation |
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.Specifications
EDA/Ectodysplasin | |
1.0 mg/mL | |
ELISA, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Mouse, Human | |
ectodysplasin A, ED1 | |
EDA | |
IgG1 | |
Protein G purified | |
Recognizes human and mouse EDA-A1 and EDA-A2. |
Renzo-2 | |
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:100 | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
1896 | |
Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title