Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDA/Ectodysplasin Antibody (Renzo-2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB001438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001438 Supplier Novus Biologicals Supplier No. NBP259772G0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen EDA/Ectodysplasin
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone Renzo-2
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA
Formulation 50 mM Sodium Borate
Gene Alias ectodysplasin A, ED1
Gene Symbols EDA
Host Species Mouse
Immunogen Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391).
Quantity 0.1 mL
Gene ID (Entrez) 1896
Test Specificity Recognizes human and mouse EDA-A1 and EDA-A2.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.