Learn More
Description
Specifications
Specifications
| Antigen | EDA2R/TNFRSF27/XEDAR |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 72 |
| Conjugate | DyLight 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human EDA2R/TNFRSF27/XEDAR (NP_068555.1; Met1-Thr138). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
