Learn More
EDAR, Rabbit anti-Human,Mouse,Rat,Canine,Chicken, Clone: EDAR12, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | EDAR |
| Applications | Western Blot, ELISA, Blocking Assay |
| Classification | Monoclonal |
| Clone | EDAR12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Surface Plasmon Resonance, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR |
| Gene Symbols | EDAR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.