Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ 48-125 Polyurethane Coated Polyester Gloves

Catalog No. 19500786 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyurethane coating on the palm and fingers provides a high level of abrasion resistance, as well as enhanced grip capabilities.

  • EN level 4 of abrasion resistance
  • Tear resistance
  • Good grip for improved productivity
  • Shows dirt contamination quickly
  • Seamless structure to ensure comfort and encourage safety
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category ll; REACH Compliant
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size 8
For Use With (Application) Inspection, General Handling, and Packaging
Glove Cuff Style Knit
Series 48-125
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.