Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Bismuth Sulfite Agar

Catalog No. ebp2069
Change view
Click to view available options
:
Bismuth Sulfite Agar; 15 x 100mm; Container: Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-P2069 Bismuth Sulfite Agar; 15 x 100mm; Container: Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P2069 Supplier Edge Biologicals Supplier No. P2069
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolation of Salmonella species especially Salmonella typhi

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Content And Storage 8°C to 12°C
Form Dehydrated
Media Type Bismuth Sulfite Agar
Product Type Agar
Formulation Peptone 10g/L, Beef extract 5g/L, Dextrose 5g/L, Disodium Phosphate 4g/L, Ferrous Sulfate 0.3g/L, Bismuth Sulfite Indicator 8g/L, Brilliant green 0.025g/L, Agar 20g/L, Deionized Water 1000mL
For Use With (Application) Isolation of Salmonella species especially Salmonella typhi
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.