Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edge Biologicals Dehydrated Culture Media: Lowenstein Jensen Agar
Click to view available options
:
Lowenstein Jensen Agar; 20 x 125mm; Container: Tube, slant
Specifications
Specifications
Form | Dehydrated |
Media Type | Lowenstein Jensen Agar |
Product Type | Agar |
For Use With (Application) | For Mycobacteria detection |
Packaging Type | Slant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction