Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edge Biologicals Dehydrated Culture Media: Mueller Hinton Agar
Click to view available options
For Use With (Application):
Test microbial susceptibility
Media Type:
Mueller Hinton Agar
Mueller Hinton Agar with Blood
Specifications
Specifications
Form | Dehydrated |
Media Type | Mueller Hinton Agar with Blood |
Product Type | Agar |
For Use With (Application) | Test microbial susceptibility |
Includes | Blood |
Packaging Type | Plate |
Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction