Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Dehydrated Culture Media: Sab Dex Agar

Catalog No. p-4901956
Click to view available options
Packaging Type:
Plate
Includes:
Chloramphenicol
Gentamicin
Media Type:
Sab Dex Agar with Chloramphenicol
Sab Dex Agar with Gentamicin
For Use With (Application):
Yeast and fungal isolation

For isolation of pathogenic Yeasts and fungi

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Description 15 x 100mm
Form Dehydrated
Media Type Sab Dex Agar with Gentamicin
Product Type Agar
For Use With (Application) Yeast and fungal isolation
Includes Gentamicin
Packaging Type Plate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.