Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edge Biologicals Dehydrated Culture Media: Tryptic Soy Broth
Click to view available options
Quantity:
4.5 mL
10 mL
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Form | Dehydrated |
| Media Type | Tryptic Soy Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dextrose 2.5g/L, Dipotassium Phosphate 2.5g/L, Deionized Water 1000mL |
| For Use With (Application) | Cultivation of fastidious and non-fastidious microorganisms |
| Packaging Type | Tube |
| Quantity | 4.5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction