Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDIL3/DEL1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP128632A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP128632A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP128632A70 Supplier Novus Biologicals Supplier No. NBP128632AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDIL3/DEL1 Polyclonal specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EDIL3/DEL1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287
Gene Symbols EDIL3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-ProtNo. O43854]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10085
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.