Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDIL3/DEL1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP128632B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128632B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128632B Supplier Novus Biologicals Supplier No. NBP128632B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDIL3/DEL1 Polyclonal specifically detects EDIL3/DEL1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EDIL3/DEL1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O43854
Gene Alias DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287
Gene Symbols EDIL3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-Prot# O43854]
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10085
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.