Learn More
Description
Specifications
Specifications
| Antigen | EDIL3/DEL1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 |
| Gene Symbols | EDIL3 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide made to an internal portion of human Del-1 (within residues 200-350). [Swiss-ProtNo. O43854] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
