Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDNRA/Endothelin R Type A Antibody, Novus Biologicals™
SDP

Catalog No. NB018259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018259 0.025 mg
NB600836 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018259 Supplier Novus Biologicals Supplier No. NB6008360.025MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 4 publications

EDNRA/Endothelin R Type A Polyclonal specifically detects EDNRA/Endothelin R Type A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen EDNRA/Endothelin R Type A
Applications Immunofluorescence, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 20-40 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with No Preservative
Gene Alias endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR
Gene Symbols EDNRA
Host Species Sheep
Immunogen Synthetic peptide corresponding to amino acids 410-422 (QEQNHNTERSSHK) of Rat Endothelin A Receptor, conjugated to KLH.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1909
Test Specificity This recognizes epitopes on the cytoplasmic side of the receptor and will not block the binding of endothelins. Therefore, the cell membrane must be permeable to the.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.