Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDNRA/Endothelin R Type A Sheep anti-Mouse, DyLight 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB600836DL5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600836DL5 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600836DL5 Supplier Novus Biologicals Supplier No. NB600836DL594

Sheep Polyclonal Antibody

EDNRA/Endothelin R Type A Polyclonal specifically detects EDNRA/Endothelin R Type A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EDNRA/Endothelin R Type A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR
Gene Symbols EDNRA
Host Species Sheep
Immunogen Synthetic peptide corresponding to amino acids 410-422 (QEQNHNTERSSHK) of Rat Endothelin A Receptor, conjugated to KLH.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1909
Test Specificity This recognizes epitopes on the cytoplasmic side of the receptor and will not block the binding of endothelins. Therefore, the cell membrane must be permeable to the.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.