Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Buffer, No Magnesium Water Hardness Buffer, Reagents

Catalog No. 01335336
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L

EDTA Buffer, No Magnesium Water Hardness Buffer, Reagent, use for Metals analysis (EDTA method). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
For Use With (Application) Metals Analysis (EDTA method)
pH 10
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Sterility Non-sterile
Synonym Water Hardness Buffer
Solubility Information Completely miscible
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.