Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AP LAB 7: Cell Division — Mitosis and Meiosis
SDPSCI_ED

Catalog No. S06885 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06885 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06885 Supplier Edvotek™ Supplier No. AP07
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Pipe cleaners (1.2 in. [3cm] long, in two colors), carbol-fuschin (Ziehl-Neelson) stain, lectin (phytohemagglutinin PHA-M from Phaseolus vulgaris), plastic bags, slides and cover slips

Requires

Colored pencils (two colors), microscope, 10 onion bulbs, jars with lids, two jars (one per treatment) for six students, sand, ethanol, glacial acetic acid (17.4M), hydrochloric acid, razor blades, forceps, scissors, scientific cleaning (Kimwipes™) wipes, coplin jars and disposable gloves

Students identify and differentiate the various stages in mitosis and meiosis.

  • Onion root tips are stained to identify the various stages and duration of mitosis
  • Meiosis and Crossing Over in Sordaria is also demonstrated in this experiment
  • Students will also have an opportunity to analyze mechanisms involved with the loss of cell cycle control in cancer

Order Info

  • Class size: 10 lab groups
  • Requires 60 minutes

Specifications

Grade 9 to 12
For Use With (Application) Students identify and differentiate various stages in mitosis and meiosis
Includes Instructions, pipe cleaners (in two colors), plastic beads, carbol-fuschin (ziehl-neelson) stain, lectin, plastic bags, slides, cover slips, sand, conical tubes, plastic cups
Time Range 60 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Cell Division Mitosis and Meiosis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.