Learn More
Description
Includes
Instructions, PCR EdvoBeads™ PLUS, PV92 LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, LyphoControl™, Universal DNA Buffer, TE Buffer, Proteinase K, UltraSpec-Agarose™, TBE Electrophoresis Buffer Powder, SYBR™Safe Stain, FlashBlue™ Stain, Microcentrifuge & PCR Tubes, Disposable Plastic Cups, Salt Packets, & a 15 mL conical tube
Requires
- Thermal cycler
- Horizontal gel electrophoresis apparatus
- Microcentrifuge
- Automatic micropipets
- UV transilluminator
- 500mL flasks
- Glassware
- All necessary safety equipment
- Ice
- Ice bucket
- Distilled or deionized water
Safe experiment for classroom use to demonstrate DNA fingerprinting.
Alu-Human DNA Typing (S66823)
- Examine students' DNA for an Alu element on Chromosome 16
- Investigate DNA patterns
- Compare Alu genotypes with those of their classmates
VNTR Human DNA Typing (S66824)
- Use PCR to fingerprint students' DNA at the D1S80 locus on Chromosome 1
- Provide an example of the variable number of tandem repeat (VNTR) polymorphism
Specifications
Specifications
| Content And Storage | Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C |
| Detection Method | Gel Electrophoresis |
| Form | Lyophilized |
| GC-Rich PCR Performance | Medium |
| PCR Method | Standard PCR |
| Reaction Speed | 60 min. |
| Label or Dye | SYBR Green, FlashBlue Stain |
| Product Type | Classroom Experiment |
| Quantity | 1 Kit |
| For Use With (Application) | Standard PCR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

