Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edwardsiella Tarda Antibody (236/128), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB162540AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162540AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162540AH Supplier Novus Biologicals Supplier No. NB10062540AF647

Mouse Monoclonal Antibody

Edwardsiella Tarda Monoclonal specifically detects Edwardsiella Tarda in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Edwardsiella Tarda
Applications ELISA
Classification Monoclonal
Clone 236/128
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias aracolobactrum anguillimortiferum, Edwardsiella anguillimortifera
Host Species Mouse
Immunogen Native
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes Edwardsiella tarda. E. tarda is a member of the family Enterobactericeae associated with freshwater ecosystems that that can cause gastoenteritis, wound infections and death due to septicaemia. It is thought that the pathogenicity of this bacteria is due to it's ability to resist phagocytic killing by the host defences. Studies show that replication of E. tarda in murine macrophages is dependant on the type III secretion system and induces an anti-apoptotic effect by up-regulating anti-apoptotic NF-kB target genes.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.