Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edwardsiella Tarda Antibody (236/128) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017492 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017492 0.025 mg
NB10062540 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017492 Supplier Novus Biologicals Supplier No. NB100625400.025MG

Mouse Monoclonal Antibody

Edwardsiella Tarda Monoclonal specifically detects Edwardsiella Tarda in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Edwardsiella Tarda
Applications ELISA
Classification Monoclonal
Clone 236/128
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Host Species Mouse
Immunogen Native
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes Edwardsiella tarda. E. tarda is a member of the family Enterobactericeae associated with freshwater ecosystems that that can cause gastoenteritis, wound infections and death due to septicaemia. It is thought that the pathogenicity of this bacteria is due to it's ability to resist phagocytic killing by the host defences. Studies show that replication of E. tarda in murine macrophages is dependant on the type III secretion system and induces an anti-apoptotic effect by up-regulating anti-apoptotic NF-kB target genes.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.