Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEA1 Antibody (6D4.1D4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP236568 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236568 0.1 mg
NBP236568SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236568 Supplier Novus Biologicals Supplier No. NBP236568
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EEA1 Monoclonal specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen EEA1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone 6D4.1D4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33212210), Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular) 2.5 ug/million cells
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q15075
Gene Alias early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Gene Symbols EEA1
Host Species Mouse
Immunogen Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075]
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endosome Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 8411
Target Species Human, Mouse, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.