Learn More
Invitrogen™ EEA1 Monoclonal Antibody (1G11), eBioscience™, Invitrogen™
Description
Description: The monoclonal antibody 1G11 recognizes human, mouse and rat early endosome antigen 1 (EEA1). EEA1 is a hydrophilic peripheral membrane protein found in the cytosol and present on early endosome membranes. EEA1 is a dimer that is localized to early endosomes via a cysteine-rich zinc-finger-like FYVE domain. EEA1 interacts with Rab5-GTP, syntaxin 6 and syntaxin 13 (SNARES). EEA1 is required for endocytic membrane docking and fusion, which allows for the recycling of receptors from the plasma membrane or their delivery to lysosomes for degradation. In neurons, early endosomes are involved in the recycling of neurotransmitter receptors. Autoantibodies to EEA1 have been found in patients with neurological deficits which result in enhanced excitatory synaptic transmission. Applications Reported: This 1G11 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, ELISA, and immunocytochemistry. Applications Tested: This 1G11 antibody has been tested by immunocytochemistry of fixed and permeabilized HeLa cells and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | EEA1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1G11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | EEA1 |
| Gene Accession No. | Q15075, Q8BL66 |
| Gene Alias | A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2 |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.