Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670159
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
01-670-159 100 μg
1 options

Catalog No. 01-670-159

Supplier: Invitrogen™ PA1063A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-063A detects early endosomal antigen 1 (EEA1) from mouse and human samples. Previous lots of this antibody have been used on hamster, canine, and rat samples. PA1-063A has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~160 kDa protein representing EEA1 from mouse brain extract. Previous lots of this antibody have been used in immunoprecipitation and immunofluorescence procedures. The PA1-063A immunogen is a synthetic peptide corresponding to residues T(1391) P S S K K P V R V C D A C F N D L Q G(1410) of human EEA1. The PA1-063A immunizing peptide (Cat. # PEP-079) is available for use in neutralization and control experiments.

Early endosomal antigen 1 (EEA1) is a 162 kDa membrane bound protein component specific to the early endosome and is essential for fusion between early endocytic vesicles. Early endosomes are cytoplasmic compartments which fuse with endocytic vesicles for redistribution of extracellular compounds to alternate destinations. Zinc-finger-like domains, reminiscent of those found in nucleic acid binding proteins, are located in the amino and carboxyl-terminal domains of EEA1. The carboxyl-terminal zinc-finger-like-domain is conserved in several other non-nuclear proteins, some of which are also involved in intracellular protein trafficking. In addition, this domain is an authentic zinc-binding domain and is critical to the intracellular localization of EEA1. Membrane association of EEA1 has been shown dependent on phosphatidyl 3-kinase activity, inhibitors of which cause EEA1 to dissociate from early endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene EEA1
Gene Accession No. Q15075, Q8BL66
Gene Alias A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2
Gene Symbols EEA1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues T(1391) P S S K K P V R V C D A C F N D L Q G(1410) of human EEA1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216238, 8411
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.