Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EED 226
GREENER_CHOICE

Catalog No. p-7249494
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective EED inhibitor

EED 226 is a potent (IC50 = 23.4 nM) and selective EED inhibitor. It inhibits PRC2 activity by binding to the H3K27me3 pocket of EED. The compound shows selectivity for PRC2 complex over protein methyltransferases, kinases, and other protein classes. In vitro, EED 226 regulates histone H3K27 methylation and PRC2 target gene expression. In vivo, it induces tumor regression in a mouse xenograft model. EED 226 is orally bioavailable.

Specifications

Chemical Name or Material N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine
CAS 2083627-02-3
Quantity 10 mg
Target EED
Molecular Formula C17H15N5O3S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.