Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eEF1A1 Antibody (23C11), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB160713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160713 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160713 Supplier Novus Biologicals Supplier No. NBP259411JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

eEF1A1 Monoclonal antibody specifically detects eEF1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen eEF1A1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 23C11
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14
Host Species Mouse
Immunogen Recombinant human eEF1A1 (1-462aa) purified from E.coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1915
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.