Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eEF1A1 Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB098863 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098863 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098863 Supplier Novus Biologicals Supplier No. NBP299359IR

Rabbit Polyclonal Antibody

eEF1A1 Polyclonal specifically detects eEF1A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen eEF1A1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human eEF1A1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1915
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.