Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eEF1A2 binding protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
| Antigen | eEF1A2 binding protein |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
eEF1A2 binding protein Polyclonal antibody specifically detects eEF1A2 binding protein in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| eEF1A2 binding protein | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp434B1231, eEF1A2 binding protein, EEF1A2-binding protein 1, EEF1A2BP1, immunoglobulin-like and fibronectin type III domain containing 1, immunoglobulin-like and fibronectin type III domain-containing protein 1, KY-interacting protein 1, KYIP1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AHSFRIRVAACPQAPGPIHLQENVPGTVTAEWEPSPDEAQDVPLHYAVFTRSSAHGPWHEAADRIYTNRFTLLGILPGHEYHFRVVA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 91156 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title