Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eEF1A2 binding protein Antibody, Novus Biologicals™
SDP

Catalog No. NB034951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034951 Supplier Novus Biologicals Supplier No. NBP287329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eEF1A2 binding protein Polyclonal specifically detects eEF1A2 binding protein in Human samples. It is validated for Western Blot.

Specifications

Antigen eEF1A2 binding protein
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias DKFZp434B1231, eEF1A2 binding protein, EEF1A2-binding protein 1, EEF1A2BP1, immunoglobulin-like and fibronectin type III domain containing 1, immunoglobulin-like and fibronectin type III domain-containing protein 1, KY-interacting protein 1, KYIP1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Human eEF1A2 binding protein
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91156
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.