Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF1B2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB087333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087333 Supplier Novus Biologicals Supplier No. NBP300341AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EEF1B2 Polyclonal specifically detects EEF1B2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EEF1B2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias EEF1B, EEF1B1, EF1B, EF-1-beta, elongation factor 1-beta, eukaryotic translation elongation factor 1 beta 1, eukaryotic translation elongation factor 1 beta 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EEF1B2 (P24534; Met1-Ile225).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1933
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.