Learn More
EEF1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 100952AP150UL

Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human EEF1B2.
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
EEF1B2 | |
Polyclonal | |
Unconjugated | |
EEF1B2 | |
EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1933, 363241, 55949 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P24534 | |
Eef1b2 | |
EEF1B2 Fusion Protein Ag0135 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.