Learn More
EEF1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | EEF1B2 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6019
![]()
|
Proteintech Group Inc
100952AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human EEF1B2.
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
| EEF1B2 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P24534 | |
| 1933, 363241, 55949 | |
| EEF1B2 Fusion Protein Ag0135 | |
| Primary | |
| -20°C | |
| EEF1B2 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
| Eef1b2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.