Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF1E1 Antibody, Novus Biologicals™
SDP

Catalog No. NB430859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB430859 25 μL
NBP189186 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB430859 Supplier Novus Biologicals Supplier No. NBP18918625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EEF1E1 Polyclonal specifically detects EEF1E1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EEF1E1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias AIMP3Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3, ARS-interacting multifunctional protein 3, Elongation factor p18, eukaryotic translation elongation factor 1 epsilon 1, eukaryotic translation elongation factor 1 epsilon-1, Multisynthase complex auxiliary component p18, P18, p18 component of aminoacyl-tRNA synthetase complex
Gene Symbols EEF1E1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LSLLEKSLGLSKGNKYSAQGERQIPVLQTNNGPSLTGLTTIAAHLVKQANKEYLLGSTAEEKAIVQQWLEYRVTQVDGHSSKNDIHTLLKDL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9521
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.