Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEN Antibody (2A9H4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252560 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252560 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252560 Supplier Novus Biologicals Supplier No. NBP252560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EEN Monoclonal specifically detects EEN in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen EEN
Applications Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
Classification Monoclonal
Clone 2A9H4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8
Gene Symbols SH3GL1
Host Species Mouse
Immunogen Purified recombinant fragment of human SH3GL1 (AA: 12-119) expressed in E. Coli.
Molecular Weight of Antigen 41.5 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6455
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.