Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEN Antibody (2A9H4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | EEN |
---|---|
Clone | 2A9H4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
EEN Monoclonal specifically detects EEN in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
EEN | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8 | |
SH3GL1 | |
IgG1 | |
Protein A or G purified |
2A9H4 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
6455 | |
Purified recombinant fragment of human SH3GL1 (AA: 12-119) expressed in E. Coli. | |
Primary | |
41.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title