Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EG-VEGF/PK1 Antibody (MM0043-2L13), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1109314A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1109314A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1109314A94 Supplier Novus Biologicals Supplier No. NB11093514AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EG-VEGF/PK1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0043-2L13
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Gene Symbols PROK1
Host Species Mouse
Immunogen human recombinant EG-VEGF protein
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84432
Test Specificity This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.