Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EG-VEGF/PK1 Antibody (MM0043-2L13) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018048 0.025 mg
NB11093514 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018048 Supplier Novus Biologicals Supplier No. NB110935140.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EG-VEGF/PK1
Applications Immunohistochemistry (Paraffin), Western Blot, ELISA
Classification Monoclonal
Clone MM0043-2L13
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1000, ELISA 1:100-1000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Gene Symbols PROK1
Host Species Mouse
Immunogen human recombinant EG-VEGF protein
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 84432
Test Specificity This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse, Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.