Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EG-VEGF/PK1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP243852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243852 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243852 Supplier Novus Biologicals Supplier No. NBP243852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects EG-VEGF/PK1 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry, Western Blotting

EG-VEGF/PK1 Polyclonal specifically detects EG-VEGF/PK1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EG-VEGF/PK1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1 000
Formulation 0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative
Gene Alias EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Gene Symbols PROK1
Host Species Rabbit
Immunogen Synthetic peptide derived from N- term domain of the human Prokineticin-1.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 84432
Reconstitution Reconstitute with distilled water to a final concentration of 1.0 mg/ml.
Target Species Human
Content And Storage Lyophilized antibody can be kept at 4C for up to 3 months and should be kept at -20C for long-term storage. To avoid freeze-thaw cycles, reconstituted antibody should be aliquoted before freezing for short-term storage (-20C) or for long-term storage (-80C). For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.