Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EG-VEGF/PK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
| Antigen | EG-VEGF/PK1 |
|---|---|
| Clone | 188608 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12091SP
![]() |
R&D Systems
MAB12091SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12091
![]() |
R&D Systems
MAB12091 |
500 μg |
Each for $643.50
|
|
|||||
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| EG-VEGF/PK1 | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P58294 | |
| 84432 | |
| Mouse myeloma cell line NS0-derived recombinant human EG-VEGF/PK1 Ala20-Phe105 Accession # P58294 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EG-VEGF/PK1 in direct ELISAs and Western blots. |
| 188608 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGVEGF, EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, PROK1, prokineticin 1 | |
| PROK1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title