Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EGCG
GREENER_CHOICESDP

Catalog No. 452450 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-245-0 Supplier R&D Systems™ Supplier No. 4524/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. β-secretase inhibitor; inhibits amyloid assembly

β-secretase (BACE1) inhibitor (IC50 = 1.6 μM); inhibits amyloid assembly. Inhibits the formation of amyloid fibrils by proteins such as Aβ, α-synuclein and huntingtin. Reduces Aβ42-induced cell death in three different cell types. Also inhibits glutamate dehydrogenase. Inhibits cell growth and selectively induces apoptosis in A431 and human pancreatic cancer cells. Also a DNMT1 inhibitor.

Chemical Identifiers

CAS 989-51-5
Molecular Formula C22H18O11

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 458.37
Purity >99%
Molecular Weight (g/mol) 458.375
Chemical Name or Material EGCG
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.