Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Antibody (008), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB306983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306983 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306983 Supplier Novus Biologicals Supplier No. NBP289582APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for ELISA

Specifications

Antigen EGF
Applications ELISA
Classification Monoclonal
Clone 008
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EGF (Accession#: NP_001954.2; Asn 971-Arg 1023) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.