Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Rabbit anti-Human, Clone: R001, Novus Biologicals™
SDP

Catalog No. NB020646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020646 500 μg
NB020645 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020646 Supplier Novus Biologicals Supplier No. NBP289581500UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EGF Monoclonal specifically detects EGF in Human samples. It is validated for Neutralization.

Specifications

Antigen EGF
Applications Neutralization
Classification Monoclonal
Clone R001
Conjugate Unconjugated
Formulation 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG
Host Species Rabbit
Immunogen Recombinant Human EGF Protein
Purification Method Protein A purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.