Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGF Rabbit anti-Human, Clone: R001, Novus Biologicals™

Rabbit Monoclonal Antibody
$572.50 - $768.00
Specifications
Antigen | EGF |
---|---|
Clone | R001 |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
EGF Monoclonal specifically detects EGF in Human samples. It is validated for Neutralization.Specifications
EGF | |
Neutralization | |
Unconjugated | |
Rabbit | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
1950 | |
IgG | |
Protein A purified |
R001 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG | |
Recombinant Human EGF Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title