Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eGFP Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB350934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350934 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350934 Supplier Novus Biologicals Supplier No. NBP306640APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eGFP Polyclonal antibody specifically detects eGFP in Nonspecies specific samples. It is validated for ELISA

Specifications

Antigen eGFP
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant eGFP (AFI57842.2; Met1-Lys239).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.