Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Antibody (h-R3 (Nimotuzumab)), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB151022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151022 Supplier Novus Biologicals Supplier No. NBP275186AF647
Add to Cart
Add to Cart

Human Monoclonal Antibody

EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen EGFR
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone h-R3 (Nimotuzumab)
Conjugate Alexa Fluor 647
Dilution Flow Cytometry, Immunohistochemistry, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Host Species Human
Immunogen Raised by humanisation of the murine antibody R3 against EGFR purified from placenta membranes and A431 tumors.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.