Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Antibody (H9B4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP234628X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234628X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234628X Supplier Novus Biologicals Supplier No. NBP234628AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EGFR Monoclonal specifically detects EGFR in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot.

Specifications

Antigen EGFR
Applications Western Blot, ELISA
Classification Monoclonal
Clone H9B4
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Gene Symbols EGFR
Host Species Mouse
Immunogen Purified EGFR from A431 cells.
Molecular Weight of Antigen 170 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 1956
Test Specificity This MAb reacts with a cytoplasmic domain of EGFR. EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, beta cellulin & epiregulin.
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.